missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 5-bromovalerate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 126460100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl 5-bromovalerate | |
5454-83-1 | |
1.3600g/mL | |
Authentic | |
Glass bottle | |
1.4620 to 1.464 | |
MFCD00000265 | |
1.36 | |
RAVVJKCSZXAIQP-UHFFFAOYSA-N | |
methyl 5-bromopentanoate | |
79557 | |
97% |
97% | |
Brown to Colorless | |
99°C | |
97% | |
C6H11BrO2 | |
Br(CH2)4CO2CH3 | |
10 g | |
methyl 5-bromovalerate, 5-bromovaleric acid methyl ester, pentanoic acid, 5-bromo-, methyl ester, methyl .omega.-bromopentanoate, 5-bromopentanoic acid methyl ester, 5-bromo-pentanoic acid methyl ester, methyl omega-bromopentanoate, methyl5-bromovalerate, valeric acid, 5-bromo-, methyl ester, methyl-5-bromovalerate | |
COC(=O)CCCCBr | |
195.06 | |
195.06 | |
Liquid |
Chemical Identifiers
5454-83-1 | |
195.06 | |
RAVVJKCSZXAIQP-UHFFFAOYSA-N | |
methyl 5-bromopentanoate |
C6H11BrO2 | |
MFCD00000265 | |
79557 | |
COC(=O)CCCCBr |
Safety and Handling
EINECSNumber : 226-709-7
TSCA : TSCA
RUO – Research Use Only