missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 5-bromosalicylate, ≥98%

$91.30 - $334.64

Chemical Identifiers

CAS 4068-76-2
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.045
MDL Number MFCD00016460
InChI Key FJYDBKPPGRZSOZ-UHFFFAOYSA-N
Synonym methyl 5-bromosalicylate, benzoic acid, 5-bromo-2-hydroxy-, methyl ester, 5-bromosalicylic acid methyl ester, methyl 5-bromo-2-hydroxybenzenecarboxylate, methyl 2-hydroxy-5-bromobenzoate, 5-bromo-2-hydroxy-benzoic acid methyl ester, methyl5-bromosalicylate, pubchem19740, acmc-1afpq, methyl-5-bromosalicylate
PubChem CID 77683
IUPAC Name methyl 5-bromo-2-hydroxybenzoate
SMILES COC(=O)C1=C(C=CC(=C1)Br)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6437206 View Documents Alfa Aesar™
H6437206
5g Each for $91.30
 
AAH6437214 View Documents Alfa Aesar™
H6437214
25g Each for $334.64
 
Specifications

Chemical Identifiers

4068-76-2
231.045
FJYDBKPPGRZSOZ-UHFFFAOYSA-N
77683
COC(=O)C1=C(C=CC(=C1)Br)O
C8H7BrO3
MFCD00016460
methyl 5-bromosalicylate, benzoic acid, 5-bromo-2-hydroxy-, methyl ester, 5-bromosalicylic acid methyl ester, methyl 5-bromo-2-hydroxybenzenecarboxylate, methyl 2-hydroxy-5-bromobenzoate, 5-bromo-2-hydroxy-benzoic acid methyl ester, methyl5-bromosalicylate, pubchem19740, acmc-1afpq, methyl-5-bromosalicylate
methyl 5-bromo-2-hydroxybenzoate

Specifications

Brown to White
5g
C8H7BrO3
3047854
FJYDBKPPGRZSOZ-UHFFFAOYSA-N
methyl 5-bromo-2-hydroxybenzoate
77683
Solid
Methyl 5-bromosalicylate
61°C to 65°C (literature)
4068-76-2
MFCD00016460
methyl 5-bromosalicylate, benzoic acid, 5-bromo-2-hydroxy-, methyl ester, 5-bromosalicylic acid methyl ester, methyl 5-bromo-2-hydroxybenzenecarboxylate, methyl 2-hydroxy-5-bromobenzoate, 5-bromo-2-hydroxy-benzoic acid methyl ester, methyl5-bromosalicylate, pubchem19740, acmc-1afpq, methyl-5-bromosalicylate
COC(=O)C1=C(C=CC(=C1)Br)O
231.045
231.05
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning