missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 5-Bromopyridine-2-carboxylate, TCI America™

$25.97 - $82.50

Chemical Identifiers

CAS 29682-15-3
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD04112493
InChI Key JEURNBCYNWNADN-UHFFFAOYSA-N
Synonym methyl 5-bromopicolinate, 5-bromopyridine-2-carboxylic acid methyl ester, methyl 5-bromo-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-pyridinecarboxylic acid methyl ester, 5-bromo-pyridine-2-carboxylic acid methyl ester, methyl5-bromopicolinate, pubchem10589, 5-bromopyridine-2-carboxylicacidmethylester, acmc-1cjr7
PubChem CID 7016458
IUPAC Name methyl 5-bromopyridine-2-carboxylate
SMILES COC(=O)C1=NC=C(C=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M26145G View Documents TCI America
M26145G
5g Each for $82.50
 
M26141G View Documents TCI America
M26141G
1g Each for $25.97
 
Specifications

Chemical Identifiers

29682-15-3
216.034
JEURNBCYNWNADN-UHFFFAOYSA-N
7016458
COC(=O)C1=NC=C(C=C1)Br
C7H6BrNO2
MFCD04112493
methyl 5-bromopicolinate, 5-bromopyridine-2-carboxylic acid methyl ester, methyl 5-bromo-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-pyridinecarboxylic acid methyl ester, 5-bromo-pyridine-2-carboxylic acid methyl ester, methyl5-bromopicolinate, pubchem10589, 5-bromopyridine-2-carboxylicacidmethylester, acmc-1cjr7
methyl 5-bromopyridine-2-carboxylate

Specifications

White-Yellow
5g
C7H6BrNO2
methyl 5-bromopicolinate, 5-bromopyridine-2-carboxylic acid methyl ester, methyl 5-bromo-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 5-bromo-, methyl ester, 5-bromo-2-pyridinecarboxylic acid methyl ester, 5-bromo-pyridine-2-carboxylic acid methyl ester, methyl5-bromopicolinate, pubchem10589, 5-bromopyridine-2-carboxylicacidmethylester, acmc-1cjr7
COC(=O)C1=NC=C(C=C1)Br
216.034
216.03
≥98.0% (GC)
100°C
29682-15-3
MFCD04112493
JEURNBCYNWNADN-UHFFFAOYSA-N
methyl 5-bromopyridine-2-carboxylate
7016458
Crystal-Powder at 20°C
Methyl 5-Bromopyridine-2-carboxylate
Safety and Handling

Safety and Handling

TSCA : No