Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 5-bromopyridine-2-carboxylate, 98+%

$82.97 - $82.97

Chemical Identifiers

Chemical Name or Material Methyl 5-bromopyridine-2-carboxylate
CAS 29682-15-3
Molecular Formula C7H6BrNO2
Formula Weight 216.03
MDL Number MFCD04112493
Synonym Methyl 5-bromopicolinate; 5-Bromopyridine-2-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2541503

  SDS
alfa aesar™
B2541503
1g Each for $82.97
Description & Specifications

Specifications

Formula Weight 216.03
Chemical Name or Material Methyl 5-bromopyridine-2-carboxylate
Assay ≥98%
Molecular Formula C7H6BrNO2
MDL Number MFCD04112493
CAS 29682-15-3
Synonym Methyl 5-bromopicolinate; 5-Bromopyridine-2-carboxylic acid methyl ester
Health Hazard 1 Warning
Melting Point 99° to 103°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Methyl 5-bromopyridine-2-carboxylate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.