missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 5-bromo-6-chloronicotinate, 98%

$31.61 - $444.14

Chemical Identifiers

CAS 78686-77-8
Molecular Formula C7H5BrClNO2
Molecular Weight (g/mol) 250.476
MDL Number MFCD03844855
InChI Key WINGWVOUOFMOJQ-UHFFFAOYSA-N
Synonym methyl 5-bromo-6-chloronicotinate, 5-bromo-6-chloro-nicotinic acid methyl ester, methyl-5-bromo-6-chloronicotinate, 3-pyridinecarboxylic acid, 5-bromo-6-chloro-, methyl ester, methyl 3-bromo-2-chloropyridine-5-carboxylate, zlchem 1175, 3-bromo-4-chloro-benzoicacidmethylester, pubchem12980, acmc-1bdei, methyl5-bromo-6-chloronicotinate
PubChem CID 2764313
IUPAC Name methyl 5-bromo-6-chloropyridine-3-carboxylate
SMILES COC(=O)C1=CC(=C(N=C1)Cl)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6475403 View Documents Alfa Aesar™
H6475403
1g Each for $31.61
 
AAH6475406 View Documents Alfa Aesar™
H6475406
5g Each for $111.80
 
AAH6475414 View Documents Alfa Aesar™
H6475414
25g Each for $444.14
 
Specifications

Chemical Identifiers

78686-77-8
250.476
WINGWVOUOFMOJQ-UHFFFAOYSA-N
2764313
COC(=O)C1=CC(=C(N=C1)Cl)Br
C7H5BrClNO2
MFCD03844855
methyl 5-bromo-6-chloronicotinate, 5-bromo-6-chloro-nicotinic acid methyl ester, methyl-5-bromo-6-chloronicotinate, 3-pyridinecarboxylic acid, 5-bromo-6-chloro-, methyl ester, methyl 3-bromo-2-chloropyridine-5-carboxylate, zlchem 1175, 3-bromo-4-chloro-benzoicacidmethylester, pubchem12980, acmc-1bdei, methyl5-bromo-6-chloronicotinate
methyl 5-bromo-6-chloropyridine-3-carboxylate

Specifications

76°C to 79°C
78686-77-8
MFCD03844855
methyl 5-bromo-6-chloronicotinate, 5-bromo-6-chloro-nicotinic acid methyl ester, methyl-5-bromo-6-chloronicotinate, 3-pyridinecarboxylic acid, 5-bromo-6-chloro-, methyl ester, methyl 3-bromo-2-chloropyridine-5-carboxylate, zlchem 1175, 3-bromo-4-chloro-benzoicacidmethylester, pubchem12980, acmc-1bdei, methyl5-bromo-6-chloronicotinate
COC(=O)C1=CC(=C(N=C1)Cl)Br
250.476
250.48
98%
1g
C7H5BrClNO2
4403950
WINGWVOUOFMOJQ-UHFFFAOYSA-N
methyl 5-bromo-6-chloropyridine-3-carboxylate
2764313
Solid
Methyl 5-bromo-6-chloronicotinate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C