Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 5-bromo-2-methoxynicotinate, 97%

$82.97 - $286.31

Chemical Identifiers

Chemical Name or Material Methyl 5-bromo-2-methoxynicotinate
CAS 122433-41-4
Molecular Formula C8H8BrNO3
Formula Weight 246.06
MDL Number MFCD08064227
Synonym 5-Bromo-2-methoxynicotinic acid methyl ester; Methyl 5-bromo-2-methoxypyridine-3-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH50066MD

 
alfa aesar™
H50066MD
250mg Each for $82.97

AAH5006603

 
alfa aesar™
H5006603
1g Each for $286.31
Description & Specifications

Specifications

Formula Weight 246.06
Chemical Name or Material Methyl 5-bromo-2-methoxynicotinate
Assay 97%
Molecular Formula C8H8BrNO3
MDL Number MFCD08064227
CAS 122433-41-4
Synonym 5-Bromo-2-methoxynicotinic acid methyl ester; Methyl 5-bromo-2-methoxypyridine-3-carboxylate
Health Hazard 1 Warning
Quantity 250mg
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.