missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 5-bromo-2-chloronicotinate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6479122
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl 5-bromo-2-chloronicotinate | |
48°C to 53°C | |
C7H5BrClNO2 | |
100 g | |
Air, light, and moisture sensitive | |
MOMQDEDQGJAKII-UHFFFAOYSA-N | |
methyl 5-bromo-2-chloropyridine-3-carboxylate | |
2763343 | |
98% |
78686-79-0 | |
>110°C (230°F) | |
MFCD03844848 | |
4394169 | |
methyl 5-bromo-2-chloronicotinate, methyl5-bromo-2-chloronicotinate, 5-bromo-2-chloro-nicotinic acid methyl ester, methyl 5-bromo-2-chloro-3-pyridinecarboxylate, 5-bromo-2-chloronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 5-bromo-2-chloro-, methyl ester, methyl 5-bromo-2-chloro-pyridine-3-carboxylate, 5-bromo-2-chloro-3-methoxycarbonyl pyridine, pubchem12966, acmc-209pf0 | |
COC(=O)C1=CC(=CN=C1Cl)Br | |
250.476 | |
250.48 | |
Solid |
Chemical Identifiers
78686-79-0 | |
250.476 | |
MOMQDEDQGJAKII-UHFFFAOYSA-N | |
methyl 5-bromo-2-chloropyridine-3-carboxylate |
C7H5BrClNO2 | |
MFCD03844848 | |
2763343 | |
COC(=O)C1=CC(=CN=C1Cl)Br |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only