missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 5-Amino-2-methylbenzoate 98.0+%, TCI America™
Supplier: TCI America M254025G
Specifications
| Methyl 5-Amino-2-methylbenzoate | |
| 42°C | |
| MFCD08752568 | |
| 5-amino-2-methyl-benzoic acid methyl ester, 5-amino-2-methylbenzoic acid methyl ester, 5-amino-2-methyl benzoic acid methyl ester, benzoic acid,5-amino-2-methyl-, methyl ester, benzoic acid, 5-amino-2-methyl-, methyl ester, 2-methyl-5-aminobenzoicacidmethylester, methyl 5-amino-o-toluate, methyl 2-methyl-5-aminobenzoate, methyl 3-amino-6-methylbenzoate, methyl 5-amino-2-methyl-benzoate | |
| COC(=O)C1=C(C)C=CC(N)=C1 | |
| 165.19 | |
| 165.19 | |
| Crystalline Powder |
| 18595-12-5 | |
| C9H11NO2 | |
| 25 g | |
| JNPZKGOLYSCSEL-UHFFFAOYSA-N | |
| methyl 5-amino-2-methylbenzoate | |
| 15049977 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 18595-12-5 | |
| 165.19 | |
| JNPZKGOLYSCSEL-UHFFFAOYSA-N | |
| 15049977 | |
| COC(=O)C1=C(C)C=CC(N)=C1 |
| C9H11NO2 | |
| MFCD08752568 | |
| 5-amino-2-methyl-benzoic acid methyl ester, 5-amino-2-methylbenzoic acid methyl ester, 5-amino-2-methyl benzoic acid methyl ester, benzoic acid,5-amino-2-methyl-, methyl ester, benzoic acid, 5-amino-2-methyl-, methyl ester, 2-methyl-5-aminobenzoicacidmethylester, methyl 5-amino-o-toluate, methyl 2-methyl-5-aminobenzoate, methyl 3-amino-6-methylbenzoate, methyl 5-amino-2-methyl-benzoate | |
| methyl 5-amino-2-methylbenzoate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator