Please login to your online account to display your discounted pricing

Methyl 5-acetylsalicylate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  339180050

 View more versions of this product

Catalog No. AC339180050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 194.19
Quantity 5g
Color White to light yellow-orange
EINECS Number 240-532-2
Melting Point 58° to 62°C
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym Methyl 2-hydroxy-5-acetylbenzoate
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Linear Formula CH3C(O)C6H3(OH)COOCH3
CAS 16475-90-4
Name Note 98%
Molecular Formula C10 H10 O4
Assay Percent Range 98%
Chemical Name or Material Methyl 5-acetylsalicylate
MDL Number MFCD00040924

Specifications

Chemical Name or Material Methyl 5-acetylsalicylate
Name Note 98%
CAS 16475-90-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10 H10 O4
Linear Formula CH3C(O)C6H3(OH)COOCH3
Formula Weight 194.19
MDL Number MFCD00040924
Synonym Methyl 2-hydroxy-5-acetylbenzoate
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents