Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 5-(4-cyanophenyl)-2-furoate, 95%

$232.31 - $232.31

Chemical Identifiers

Chemical Name or Material Methyl 5-(4-cyanophenyl)-2-furoate
CAS 834884-75-2
Molecular Formula C13H9NO3
Formula Weight 227.22g/mol
MDL Number MFCD06797401
Synonym 5-(4-Cyanophenyl)-2-furoic acid methyl ester; Methyl 5-(4-cyanophenyl)furan-2-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5174706

 
alfa aesar™
H51747-06
Each for $232.31
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material Methyl 5-(4-cyanophenyl)-2-furoate
CAS 834884-75-2
Assay 95%
Molecular Formula C13H9NO3
Formula Weight 227.22g/mol
MDL Number MFCD06797401
Synonym 5-(4-Cyanophenyl)-2-furoic acid methyl ester; Methyl 5-(4-cyanophenyl)furan-2-carboxylate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.