missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4H-furo[3,2-b]pyrrole-5-carboxylate, 97%, Maybridge

$741.81 - $741.81

Chemical Identifiers

CAS 77484-99-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H7NO3
Formula Weight 165.15
MDL Number MFCD06797487
Chemical Name or Material Methyl 4H-furo[3, 2-b]pyrrole-5-carboxylate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC39222EA SDS Fisher Scientific Company
CC39222EA
Amber Glass Bottle 10g Each for $741.81
Specifications Description & Specifications

Chemical Identifiers

77484-99-2
100.0
C8H7NO3
MFCD06797487
97%
96.5
97%
165.15
Methyl 4H-furo[3, 2-b]pyrrole-5-carboxylate

Specifications

165.15
100.0
77484-99-2
96.5
97%
97%
10g
C8H7NO3
MFCD06797487
Amber Glass Bottle
Methyl 4H-furo[3, 2-b]pyrrole-5-carboxylate

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning