missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-vinylbenzoate, 97%

$205.11 - $727.88

Chemical Identifiers

CAS 1076-96-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00017218
InChI Key NUMHUJZXKZKUBN-UHFFFAOYSA-N
Synonym methyl 4-vinylbenzoate, methyl4-vinylbenzoate, methyl p-vinylbenzoate, methyl-4-vinylbenzoate, benzoic acid, 4-ethenyl-, methyl ester, benzoic acid, p-vinyl-, methyl ester, 4-vinyl benzoic acid methyl ester, acmc-1brgo, methyl 4-vinylbenzoate, 4-vinyl-benzoic acid methyl ester
PubChem CID 583124
IUPAC Name methyl 4-ethenylbenzoate
SMILES COC(=O)C1=CC=C(C=C1)C=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6005703 View Documents Alfa Aesar™
H6005703
1g Each for $205.11
 
AAH6005706 View Documents Alfa Aesar™
H6005706
5g Each for $727.88
 
Description

Description

Applications
Methyl 4-vinylbenzoate is used as a fluid component for enhancing ink stability in printing and also in preparing vinyl substituted beta-diketones. Further, it is used as an organic intermediate for pharmaceuticals.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

1076-96-6
162.188
NUMHUJZXKZKUBN-UHFFFAOYSA-N
583124
COC(=O)C1=CC=C(C=C1)C=C
C10H10O2
MFCD00017218
methyl 4-vinylbenzoate, methyl4-vinylbenzoate, methyl p-vinylbenzoate, methyl-4-vinylbenzoate, benzoic acid, 4-ethenyl-, methyl ester, benzoic acid, p-vinyl-, methyl ester, 4-vinyl benzoic acid methyl ester, acmc-1brgo, methyl 4-vinylbenzoate, 4-vinyl-benzoic acid methyl ester
methyl 4-ethenylbenzoate

Specifications

32°C to 37°C
1076-96-6
MFCD00017218
NUMHUJZXKZKUBN-UHFFFAOYSA-N
methyl 4-ethenylbenzoate
583124
97%
1g
C10H10O2
methyl 4-vinylbenzoate, methyl4-vinylbenzoate, methyl p-vinylbenzoate, methyl-4-vinylbenzoate, benzoic acid, 4-ethenyl-, methyl ester, benzoic acid, p-vinyl-, methyl ester, 4-vinyl benzoic acid methyl ester, acmc-1brgo, methyl 4-vinylbenzoate, 4-vinyl-benzoic acid methyl ester
COC(=O)C1=CC=C(C=C1)C=C
162.188
162.19
Methyl 4-vinylbenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning