missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-(trifluoromethyl)benzoate, 99%, ACROS Organics™

Chemical Identifiers

CAS 2967-66-0
Assay Percent Range 99%
Molecular Formula C9H7F3O2
Linear Formula CF3C6H4COCH3
Formula Weight 204.14
MDL Number MFCD00042324
Chemical Name or Material Methyl 4-(trifluoromethyl)benzoate, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC305970050 SDS Thermo Fisher Scientific
305970050
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC305970010 SDS Thermo Fisher Scientific
AC305970010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2967-66-0
C9H7F3O2
204.14
Methyl 4-(trifluoromethyl)benzoate, 99%
99%
CF3C6H4COCH3
MFCD00042324
≥98.5%

Specifications

Clear colorless to very pale yellow
2967-66-0
Authentic
Liquid
C9H7F3O2
204.14
Methyl 4-(trifluoromethyl)benzoate, 99%
5g
1.26g/mL
1.4500 to 1.4520 (20°C, 589nm)
99%
CF3C6H4COCH3
MFCD00042324
≥98.5%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.