Please login to your online account to display your discounted pricing

Methyl 4-(trifluoromethyl)benzoate, 99%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC305970010

 View more versions of this product

Catalog No. AC305970010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥98.5%
Formula Weight 204.14
Quantity 1g
Color Clear colorless to very pale yellow
Density 1.26g/mL
Refractive Index 1.4500 to 1.4520 (20°C, 589nm)
Chemical Name or Material Methyl 4-(trifluoromethyl)benzoate
Infrared Spectrum Authentic
Name Note 99%
Linear Formula CF3C6H4COCH3
MDL Number MFCD00042324
CAS 2967-66-0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99%
Molecular Formula C9H7F3O2

Specifications

Chemical Name or Material Methyl 4-(trifluoromethyl)benzoate
Name Note 99%
CAS 2967-66-0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H7F3O2
Linear Formula CF3C6H4COCH3
Formula Weight 204.14
MDL Number MFCD00042324
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents