Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-(trifluoromethoxy)phenyl sulfone, 98+%

$225.69 - $225.69

Chemical Identifiers

Chemical Name or Material Methyl 4-(trifluoromethoxy)phenyl sulfone
CAS 87750-51-4
Molecular Formula C8H7F3O3S
Formula Weight 240.20g/mol
MDL Number MFCD00792426
Beilstein 7924039
Synonym 1-Methanesulfonyl-4-(trifluoromethoxy)benzene; 1-Methylsulfonyl-4-(trifluoromethoxy)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1497806

 
alfa aesar™
L14978-06
5g Each for $225.69
Description & Specifications

Specifications

Melting Point 65° to 69°C
Quantity 5g
Chemical Name or Material Methyl 4-(trifluoromethoxy)phenyl sulfone
CAS 87750-51-4
Assay ≥98%
Molecular Formula C8H7F3O3S
Formula Weight 240.20g/mol
MDL Number MFCD00792426
Beilstein 7924039
Synonym 1-Methanesulfonyl-4-(trifluoromethoxy)benzene; 1-Methylsulfonyl-4-(trifluoromethoxy)benzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.