Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-tert-butylphenylacetate, 97%

$957.43 - $957.43

Chemical Identifiers

Chemical Name or Material Methyl 4-tert-butylphenylacetate
CAS 3549-23-3
Molecular Formula C13H18O2
Formula Weight 206.29
MDL Number MFCD00051913
Beilstein 2502880
Synonym 4-tert-Butylphenylacetic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1242122

  SDS
alfa aesar™
A1242122
100g Each for $957.43
Description & Specifications

Specifications

Formula Weight 206.29
Chemical Name or Material Methyl 4-tert-butylphenylacetate
TSCA TSCA
Assay 97%
Molecular Formula C13H18O2
MDL Number MFCD00051913
CAS 3549-23-3
Beilstein 2502880
Synonym 4-tert-Butylphenylacetic acid methyl ester
Boiling Point 149° to 151°C (30mmHg)
Quantity 100g
Flash Point >100°C (212°F)