missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 4-tert-butylphenylacetate, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1242114
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl 4-tert-butylphenylacetate | |
0.994 | |
>100°C (212°F) | |
1.4995 | |
25 g | |
methyl p-tert-butylphenylacetate, methyl 4-tert-butylphenylacetate, methyl 2-4-tert-butyl phenyl acetate, 4-tert-butylphenylacetic acid methyl ester, benzeneacetic acid, 4-1,1-dimethylethyl-, methyl ester, unii-8a4h5qo9oj, methyl 2-4-tert-butylphenyl acetate, 8a4h5qo9oj, methyl 4-1,1-dimethylethyl benzeneacetate, acetic acid, p-tert-butylphenyl-, methyl ester | |
COC(=O)CC1=CC=C(C=C1)C(C)(C)C | |
206.29 | |
206.29 |
3549-23-3 | |
149°C to 151°C (30 mmHg) | |
C13H18O2 | |
MFCD00051913 | |
2502880 | |
HXVTYMWVMVKVTF-UHFFFAOYSA-N | |
methyl 2-(4-tert-butylphenyl)acetate | |
605629 | |
97% |
Chemical Identifiers
3549-23-3 | |
206.29 | |
HXVTYMWVMVKVTF-UHFFFAOYSA-N | |
605629 | |
COC(=O)CC1=CC=C(C=C1)C(C)(C)C |
C13H18O2 | |
MFCD00051913 | |
methyl p-tert-butylphenylacetate, methyl 4-tert-butylphenylacetate, methyl 2-4-tert-butyl phenyl acetate, 4-tert-butylphenylacetic acid methyl ester, benzeneacetic acid, 4-1,1-dimethylethyl-, methyl ester, unii-8a4h5qo9oj, methyl 2-4-tert-butylphenyl acetate, 8a4h5qo9oj, methyl 4-1,1-dimethylethyl benzeneacetate, acetic acid, p-tert-butylphenyl-, methyl ester | |
methyl 2-(4-tert-butylphenyl)acetate |
Safety and Handling
EINECSNumber : 222-602-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only