missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-Piperidinecarboxylate, TCI America™

$35.14 - $106.95

Chemical Identifiers

CAS 2971-79-1
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD00190578
InChI Key RZVWBASHHLFBJF-UHFFFAOYSA-N
Synonym methyl isonipecotate, methyl 4-piperidinecarboxylate, 4-piperidinecarboxylic acid, methyl ester, piperidine-4-carboxylic acid methyl ester, 4-piperidinecarboxylic acid methyl ester, isonipecotic acid methyl ester, methylpiperidine-4-carboxylate, methylisonipecotate, methyl piperidin-4-carboxylate, acmc-20aiv4
PubChem CID 424914
IUPAC Name methyl piperidine-4-carboxylate
SMILES COC(=O)C1CCNCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M207525G View Documents TCI America
M207525G
25g Each for $106.95
 
M20755G View Documents TCI America
M20755G
5g Each for $35.14
 
Specifications

Chemical Identifiers

2971-79-1
143.186
RZVWBASHHLFBJF-UHFFFAOYSA-N
424914
COC(=O)C1CCNCC1
C7H13NO2
MFCD00190578
methyl isonipecotate, methyl 4-piperidinecarboxylate, 4-piperidinecarboxylic acid, methyl ester, piperidine-4-carboxylic acid methyl ester, 4-piperidinecarboxylic acid methyl ester, isonipecotic acid methyl ester, methylpiperidine-4-carboxylate, methylisonipecotate, methyl piperidin-4-carboxylate, acmc-20aiv4
methyl piperidine-4-carboxylate

Specifications

93°C
25g
C7H13NO2
methyl isonipecotate, methyl 4-piperidinecarboxylate, 4-piperidinecarboxylic acid, methyl ester, piperidine-4-carboxylic acid methyl ester, 4-piperidinecarboxylic acid methyl ester, isonipecotic acid methyl ester, methylpiperidine-4-carboxylate, methylisonipecotate, methyl piperidin-4-carboxylate, acmc-20aiv4
COC(=O)C1CCNCC1
143.186
143.19
≥98.0% (T)
Yellow
2971-79-1
MFCD00190578
RZVWBASHHLFBJF-UHFFFAOYSA-N
methyl piperidine-4-carboxylate
424914
Clear Liquid at 20°C
Methyl 4-Piperidinecarboxylate
Safety and Handling

Safety and Handling

TSCA : No