missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-(piperidin-1-ylmethyl)benzoate 97+%, Maybridge

Chemical Identifiers

Assay Percent Range 97%
Molecular Formula C14H19NO2
Formula Weight 233.31
Chemical Name or Material Methyl 4-(piperidin-1-ylmethyl)benzoate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC45522DA SDS Fisher Scientific Company
CC45522DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

97%
233.31
97%
C14H19NO2
Methyl 4-(piperidin-1-ylmethyl)benzoate

Specifications

233.31
C14H19NO2
Amber Glass Bottle
Methyl 4-(piperidin-1-ylmethyl)benzoate
1g
68453-37-2
97%
97%