missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-(Phenylcarbamoyl)benzoate, TCI America™

$105.42 - $348.34

Chemical Identifiers

CAS 3814-10-6
Molecular Formula C15H13NO3
Molecular Weight (g/mol) 255.27
MDL Number MFCD00157832
InChI Key XTAGEOVGQDJRIC-UHFFFAOYSA-N
Synonym 4-(Phenylcarbamoyl)benzoic Acid Methyl Ester
PubChem CID 4682178
IUPAC Name methyl 4-(phenylcarbamoyl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M26371G View Documents TCI America
M26371G
1g Each for $105.42
 
M26375G View Documents TCI America
M26375G
5g Each for $348.34
 
Specifications

Chemical Identifiers

3814-10-6
255.27
XTAGEOVGQDJRIC-UHFFFAOYSA-N
4682178
COC(=O)C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2
C15H13NO3
MFCD00157832
4-(Phenylcarbamoyl)benzoic Acid Methyl Ester
methyl 4-(phenylcarbamoyl)benzoate

Specifications

White
1g
C15H13NO3
4-(Phenylcarbamoyl)benzoic Acid Methyl Ester
COC(=O)C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2
255.27
255.27
≥98.0% (GC)
194°C
3814-10-6
MFCD00157832
XTAGEOVGQDJRIC-UHFFFAOYSA-N
methyl 4-(phenylcarbamoyl)benzoate
4682178
Crystal-Powder at 20°C
Methyl 4-(Phenylcarbamoyl)benzoate
Safety and Handling

Safety and Handling

TSCA : No