missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 4-(Phenylcarbamoyl)benzoate 98.0+%, TCI America™
Supplier: TCI America M26371G
Specifications
Methyl 4-(Phenylcarbamoyl)benzoate | |
194°C | |
C15H13NO3 | |
1 g | |
XTAGEOVGQDJRIC-UHFFFAOYSA-N | |
methyl 4-(phenylcarbamoyl)benzoate | |
4682178 | |
≥98.0% (GC) |
3814-10-6 | |
White | |
MFCD00157832 | |
4-(Phenylcarbamoyl)benzoic Acid Methyl Ester | |
COC(=O)C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2 | |
255.27 | |
255.27 | |
Crystalline Powder |
Chemical Identifiers
3814-10-6 | |
255.27 | |
XTAGEOVGQDJRIC-UHFFFAOYSA-N | |
4682178 | |
COC(=O)C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2 |
C15H13NO3 | |
MFCD00157832 | |
4-(Phenylcarbamoyl)benzoic Acid Methyl Ester | |
methyl 4-(phenylcarbamoyl)benzoate |
Safety and Handling
TSCA : No