Please login to your online account to display your discounted pricing

Methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
CAS 237385-98-7
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C13H9F3O2S
Formula Weight 286.27
MDL Number MFCD00110251
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW02756EA

  SDS
fisher scientific company
SEW02756EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 286.27
Quantity 10g
Assay 95%
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Chemical Name or Material Methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
CAS 237385-98-7
MDL Number MFCD00110251
Molecular Formula C13H9F3O2S