Please login to your online account to display your discounted pricing

Methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate, 95%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SEW02756EA

 View more versions of this product

Catalog No. SEW02756EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 286.27
Quantity 10g
Assay 95%
CAS Min % 95.0
Chemical Name or Material Methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C13H9F3O2S
MDL Number MFCD00110251
CAS 237385-98-7

Specifications

Chemical Name or Material Methyl 4-phenyl-5-(trifluoromethyl)thiophene-2-carboxylate
CAS 237385-98-7
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C13H9F3O2S
Formula Weight 286.27
MDL Number MFCD00110251