missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-Oxopiperidine-3-carboxylate Hydrochloride, TCI America™

$244.45 - $733.34

Chemical Identifiers

CAS 71486-53-8
Molecular Formula C7H12ClNO3
Molecular Weight (g/mol) 193.627
MDL Number MFCD00012769
InChI Key NMAACQILAGCQPR-UHFFFAOYSA-N
Synonym methyl 4-oxopiperidine-3-carboxylate hydrochloride, methyl 4-oxo-3-piperidinecarboxylate hydrochloride, 3-methoxycarbonyl-4-piperidone hydrochloride, methyl 4-oxo-3-piperidinecarboxylate hcl, 3-piperidinecarboxylic acid, 4-oxo-, methyl ester, hydrochloride, 3-methoxycarbonyl-4-oxopiperidinium chloride, methyl 4-oxopiperidine-3-carboxylate, chloride, 4-piperidine carboxylic acid methyl ketone 3-hydrochloride, pubchem12958, methyl 4-oxopiperidine-3-carboxylate hcl
PubChem CID 2724028
IUPAC Name methyl 4-oxopiperidine-3-carboxylate;hydrochloride
SMILES COC(=O)C1CNCCC1=O.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M285225G View Documents TCI America
M285225G
25g Each for $733.34
 
M28525G View Documents TCI America
M28525G
5g Each for $244.45
 
Specifications

Chemical Identifiers

71486-53-8
193.627
NMAACQILAGCQPR-UHFFFAOYSA-N
2724028
COC(=O)C1CNCCC1=O.Cl
C7H12ClNO3
MFCD00012769
methyl 4-oxopiperidine-3-carboxylate hydrochloride, methyl 4-oxo-3-piperidinecarboxylate hydrochloride, 3-methoxycarbonyl-4-piperidone hydrochloride, methyl 4-oxo-3-piperidinecarboxylate hcl, 3-piperidinecarboxylic acid, 4-oxo-, methyl ester, hydrochloride, 3-methoxycarbonyl-4-oxopiperidinium chloride, methyl 4-oxopiperidine-3-carboxylate, chloride, 4-piperidine carboxylic acid methyl ketone 3-hydrochloride, pubchem12958, methyl 4-oxopiperidine-3-carboxylate hcl
methyl 4-oxopiperidine-3-carboxylate;hydrochloride

Specifications

White-Yellow
25g
C7H12ClNO3
methyl 4-oxopiperidine-3-carboxylate hydrochloride, methyl 4-oxo-3-piperidinecarboxylate hydrochloride, 3-methoxycarbonyl-4-piperidone hydrochloride, methyl 4-oxo-3-piperidinecarboxylate hcl, 3-piperidinecarboxylic acid, 4-oxo-, methyl ester, hydrochloride, 3-methoxycarbonyl-4-oxopiperidinium chloride, methyl 4-oxopiperidine-3-carboxylate, chloride, 4-piperidine carboxylic acid methyl ketone 3-hydrochloride, pubchem12958, methyl 4-oxopiperidine-3-carboxylate hcl
COC(=O)C1CNCCC1=O.Cl
193.627
193.63
≥98.0% (HPLC,N)
177°C
71486-53-8
MFCD00012769
NMAACQILAGCQPR-UHFFFAOYSA-N
methyl 4-oxopiperidine-3-carboxylate;hydrochloride
2724028
Crystal-Powder at 20°C
Methyl 4-Oxopiperidine-3-carboxylate Hydrochloride
Safety and Handling

Safety and Handling

TSCA : No