Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-oxopiperidine-3-carboxylate hydrochloride, 95%

$60.18 - $600.32

Chemical Identifiers

Chemical Name or Material Methyl 4-oxopiperidine-3-carboxylate hydrochloride
CAS 71486-53-8
Molecular Formula C7H11NO3·HCl
Formula Weight 193.63
MDL Number MFCD00012769
Beilstein 4273241
Synonym 3-Methoxycarbonyl-4-piperidone hydrochloride; 4-Oxopiperidine-3-carboxylic acid methyl ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2731803

  SDS
alfa aesar™
H2731803
1g Each for $60.18

AAH2731806

  SDS
alfa aesar™
H2731806
5g Each for $175.47

AAH2731814

  SDS
alfa aesar™
H2731814
25g Each for $600.32
Description & Specifications

Specifications

Formula Weight 193.63
Health Hazard 1 Warning
Chemical Name or Material Methyl 4-oxopiperidine-3-carboxylate hydrochloride
Assay 95%
Molecular Formula C7H11NO3·HCl
MDL Number MFCD00012769
CAS 71486-53-8
Beilstein 4273241
Synonym 3-Methoxycarbonyl-4-piperidone hydrochloride; 4-Oxopiperidine-3-carboxylic acid methyl ester hydrochloride
Melting Point ≈175°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.