Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-n-butoxybenzoate, 98%

$557.22 - $557.22

Chemical Identifiers

Chemical Name or Material Methyl 4-n-butoxybenzoate
CAS 4906-25-6
Molecular Formula C12H16O3
Formula Weight 208.26
MDL Number MFCD00070801
Beilstein 2099767
Synonym 4-n-Butoxybenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1761630

 
alfa aesar™
A1761630
250g Each for $557.22
Description & Specifications

Specifications

Formula Weight 208.26
Chemical Name or Material Methyl 4-n-butoxybenzoate
Assay 98%
Molecular Formula C12H16O3
MDL Number MFCD00070801
CAS 4906-25-6
Beilstein 2099767
Synonym 4-n-Butoxybenzoic acid methyl ester
Boiling Point 188° to 190°C (30mmHg)
Quantity 250g