Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-morpholineacetate, 99%

$80.76 - $849.81

Chemical Identifiers

Chemical Name or Material Methyl 4-morpholineacetate
CAS 35855-10-8
Molecular Formula C7H13O3N
Formula Weight 159.19
MDL Number MFCD00216973
Synonym 4-Morpholineacetic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2184206

  SDS
alfa aesar™
B2184206
5g Each for $80.76

AAB2184214

  SDS
alfa aesar™
B2184214
25g Each for $272.73

AAB2184222

  SDS
alfa aesar™
B2184222
100g Each for $849.81
Description & Specifications

Specifications

Formula Weight 159.19
Chemical Name or Material Methyl 4-morpholineacetate
Assay 99%
Molecular Formula C7H13O3N
MDL Number MFCD00216973
CAS 35855-10-8
Synonym 4-Morpholineacetic acid methyl ester
Health Hazard 1 Warning
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.