missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-(methylthio)butyrate, 96%

Manufacturer:  Alfa Aesar™ L1100006

 View more versions of this product

Catalog No. AAL1100006



Specifications

Specifications

Methyl 4-(methylthio)butyrate
114°C to 115°C (40mmHg)
53053-51-3
MFCD00009849
HQDLZISNHUMXEA-UHFFFAOYSA-N
methyl 4-methylsulfanylbutanoate
65425
96%
72°C (161°F)
5g
C6H12O2S
methyl 4-methylthio butyrate, methyl 4-methylthio butanoate, methyl 4-methylsulfanyl butanoate, butanoic acid, 4-methylthio-, methyl ester, fema no. 3412, methyl gamma-methylthio butyrate, methyl gamma-methylmercapto butyrate, acmc-20akgs, methyl-4-methylthio butanoat, methyl 4-methyl thio butyrate
COC(=O)CCCSC
148.22
148.23

Chemical Identifiers

53053-51-3
148.22
HQDLZISNHUMXEA-UHFFFAOYSA-N
65425
COC(=O)CCCSC
C6H12O2S
MFCD00009849
methyl 4-methylthio butyrate, methyl 4-methylthio butanoate, methyl 4-methylsulfanyl butanoate, butanoic acid, 4-methylthio-, methyl ester, fema no. 3412, methyl gamma-methylthio butyrate, methyl gamma-methylmercapto butyrate, acmc-20akgs, methyl-4-methylthio butanoat, methyl 4-methyl thio butyrate
methyl 4-methylsulfanylbutanoate
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents