Learn More
Methyl 4-methylsalicylate, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6479314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl 4-methylsalicylate | |
27°C to 28°C | |
1.147 g/mL | |
C9H10O3 | |
MFCD00020130 | |
2690247 | |
UITFCFWKYAOJEJ-UHFFFAOYSA-N | |
methyl 2-hydroxy-4-methylbenzoate | |
78400 | |
≥98% |
4670-56-8 | |
Brown to Yellow | |
242°C (literature) | |
1.537 | |
25 g | |
methyl 4-methylsalicylate, methyl-4-methylsalicylate, benzoic acid, 2-hydroxy-4-methyl-, methyl ester, 4-methylsalicylic acid methyl ester, methyl-4-methyl salicylate, methyl m-cresotate, 2-hydroxy-4-methylbenzoic acid methyl ester, methyl 2-hydroxy-4-methyl-benzoate, 2-hydroxy-4-methyl-benzoic acid methyl ester, methyl methylsalicylate | |
CC1=CC(=C(C=C1)C(=O)OC)O | |
166.176 | |
166.18 | |
Low Melting Solid |
Chemical Identifiers
4670-56-8 | |
166.176 | |
UITFCFWKYAOJEJ-UHFFFAOYSA-N | |
78400 | |
CC1=CC(=C(C=C1)C(=O)OC)O |
C9H10O3 | |
MFCD00020130 | |
methyl 4-methylsalicylate, methyl-4-methylsalicylate, benzoic acid, 2-hydroxy-4-methyl-, methyl ester, 4-methylsalicylic acid methyl ester, methyl-4-methyl salicylate, methyl m-cresotate, 2-hydroxy-4-methylbenzoic acid methyl ester, methyl 2-hydroxy-4-methyl-benzoate, 2-hydroxy-4-methyl-benzoic acid methyl ester, methyl methylsalicylate | |
methyl 2-hydroxy-4-methylbenzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 225-117-6
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only