missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-Methylcinnamate, TCI America™

$111.53 - $330.00

Chemical Identifiers

CAS 7560-43-2
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00460746
InChI Key WLJBRXRCJNSDHT-UHFFFAOYSA-N
Synonym 4-Methylcinnamic Acid Methyl Ester
PubChem CID 589472
IUPAC Name methyl 3-(4-methylphenyl)prop-2-enoate
SMILES CC1=CC=C(C=C1)C=CC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M28251G View Documents TCI America
M28251G
1g Each for $111.53
 
M28255G View Documents TCI America
M28255G
5g Each for $330.00
 
Specifications

Chemical Identifiers

7560-43-2
176.215
WLJBRXRCJNSDHT-UHFFFAOYSA-N
589472
CC1=CC=C(C=C1)C=CC(=O)OC
C11H12O2
MFCD00460746
4-Methylcinnamic Acid Methyl Ester
methyl 3-(4-methylphenyl)prop-2-enoate

Specifications

165°C
58°C
7560-43-2
MFCD00460746
WLJBRXRCJNSDHT-UHFFFAOYSA-N
methyl 3-(4-methylphenyl)prop-2-enoate
589472
Crystal-Powder at 20°C
Methyl 4-Methylcinnamate
White
1g
C11H12O2
4-Methylcinnamic Acid Methyl Ester
CC1=CC=C(C=C1)C=CC(=O)OC
176.215
176.22
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No