missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4'-methylbiphenyl-4-carboxylate, 95%

$57.15 - $227.38

Chemical Identifiers

CAS 49742-56-5
Molecular Formula C15H14O2
Formula Weight 226.27
MDL Number MFCD05979484
Synonym 4'-Methylbiphenyl-4-carboxylic acid methyl ester
Chemical Name or Material Methyl 4'-methylbiphenyl-4-carboxylate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5191103 SDS Alfa Aesar™
H5191103
1g Each for $57.15
AAH5191106 SDS Alfa Aesar™
H5191106
5g Each for $227.38
Specifications Description & Specifications

Chemical Identifiers

49742-56-5
226.27
4'-Methylbiphenyl-4-carboxylic acid methyl ester
C15H14O2
MFCD05979484
Methyl 4'-methylbiphenyl-4-carboxylate

Specifications

49742-56-5
226.27
4'-Methylbiphenyl-4-carboxylic acid methyl ester
Methyl 4'-methylbiphenyl-4-carboxylate
C15H14O2
MFCD05979484
1g
95%

Safety and Handling

TSCA : TSCA