missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 4-Methyl-3-nitrobenzoate 98.0+%, TCI America™
Supplier: TCI America M214625G
Specifications
Methyl 4-Methyl-3-nitrobenzoate | |
51°C | |
C9H9NO4 | |
25 g | |
YFPBHPCMYFCRKS-UHFFFAOYSA-N | |
methyl 4-methyl-3-nitrobenzoate | |
81816 | |
≥98.0% (GC) |
7356-11-8 | |
Yellow | |
MFCD00130054 | |
methyl 3-nitro-4-methylbenzoate, 4-methyl-3-nitrobenzoic acid methyl ester, benzoic acid, 4-methyl-3-nitro-, methyl ester, methyl 3-nitro-p-toluate, 4-methyl-3-nitro-benzoic acid methyl ester, pubchem10934, acmc-1bfci, ksc497g4t, methyl4-methyl-3-nitrobenzoate, 4-methoxycarbonyl-2-nitrotoluene | |
CC1=C(C=C(C=C1)C(=O)OC)[N+](=O)[O-] | |
195.17 | |
195.17 | |
Crystalline Powder |
Chemical Identifiers
7356-11-8 | |
195.17 | |
YFPBHPCMYFCRKS-UHFFFAOYSA-N | |
81816 | |
CC1=C(C=C(C=C1)C(=O)OC)[N+](=O)[O-] |
C9H9NO4 | |
MFCD00130054 | |
methyl 3-nitro-4-methylbenzoate, 4-methyl-3-nitrobenzoic acid methyl ester, benzoic acid, 4-methyl-3-nitro-, methyl ester, methyl 3-nitro-p-toluate, 4-methyl-3-nitro-benzoic acid methyl ester, pubchem10934, acmc-1bfci, ksc497g4t, methyl4-methyl-3-nitrobenzoate, 4-methoxycarbonyl-2-nitrotoluene | |
methyl 4-methyl-3-nitrobenzoate |
Safety and Handling
TSCA : Yes