Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-methyl-3-nitrobenzoate, 99%

$63.57 - $849.69

Chemical Identifiers

Chemical Name or Material Methyl 4-methyl-3-nitrobenzoate
CAS 7356-11-8
Molecular Formula C9H9NO4
Formula Weight 195.18
MDL Number MFCD00130054
Synonym 4-Methyl-3-nitrobenzoic acid methyl ester; Methyl 3-nitro-p-toluate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2529906

 
alfa aesar™
B2529906
5g Each for $63.57

AAB2529922

 
alfa aesar™
B2529922
100g Each for $849.69
Description & Specifications

Specifications

Formula Weight 195.18
Chemical Name or Material Methyl 4-methyl-3-nitrobenzoate
TSCA TSCA
Assay 99%
Molecular Formula C9H9NO4
MDL Number MFCD00130054
CAS 7356-11-8
Synonym 4-Methyl-3-nitrobenzoic acid methyl ester; Methyl 3-nitro-p-toluate
Melting Point 49° to 53°C
Quantity 5g

Applications
Methyl 4-methyl-3-nitrobenzoate is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.