Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-methyl-2-pentenoate, 97%

Chemical Identifiers

Chemical Name or Material Methyl 4-methyl-2-pentenoate
CAS 50652-78-3
Molecular Formula C7H12O2
Formula Weight 128.17
MDL Number MFCD00137612
UN Number UN3272
Beilstein 1744872
Synonym 4-Methyl-2-pentenoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2019103

 
alfa aesar™
L2019103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2019114

 
alfa aesar™
L2019114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 128.17
Chemical Name or Material Methyl 4-methyl-2-pentenoate
MDL Number MFCD00137612
CAS 50652-78-3
UN Number UN3272
Synonym 4-Methyl-2-pentenoic acid methyl ester
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Molecular Formula C7H12O2
Beilstein 1744872
Health Hazard 1 Warning
Boiling Point 74° to 75°C (46mmHg)
Quantity 1g
Flash Point 47°C (116°F)
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

Applications
It is employed in the synthesis and reaction of α,β-unsaturated α-nitrocarboxylic esters.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.