Please login to your online account to display your discounted pricing

Methyl 4-methoxyphenylacetate, 97%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  189831000

 View more versions of this product

Catalog No. AC189831000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥96.0%
Quantity 100g
Color Clear colorless to yellow
Beilstein 10, 191
Chemical Name or Material Methyl 4-methoxyphenylacetate
EINECS Number 245-886-1
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
Refractive Index 1.5150 to 1.5170 (20°C, 589nm)
Assay Percent Range 97%
Molecular Formula C10H12O3
Linear Formula CH3OC6H4CH2CO2CH3
MDL Number MFCD00008454
CAS 23786-14-3
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Formula Weight 180.20
Density 1.1170g/mL

Specifications

Chemical Name or Material Methyl 4-methoxyphenylacetate
Name Note 97%
CAS 23786-14-3
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H12O3
Linear Formula CH3OC6H4CH2CO2CH3
Formula Weight 180.20
MDL Number MFCD00008454
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents