Please login to your online account to display your discounted pricing

Methyl 4-methoxyphenylacetate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 4-methoxyphenylacetate
Name Note 97%
CAS 23786-14-3
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H12O3
Linear Formula CH3OC6H4CH2CO2CH3
Formula Weight 180.20
MDL Number MFCD00008454
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC189831000

  SDS
thermo fisher scientific
189831000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥96.0%
Quantity 100g
Color Clear colorless to yellow
Beilstein 10, 191
Chemical Name or Material Methyl 4-methoxyphenylacetate
Health Hazard 1 GHS Signal Word: Warning
CAS 23786-14-3
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
MDL Number MFCD00008454
Packaging Glass bottle
EINECS Number 245-886-1
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Molecular Formula C10H12O3
Assay Percent Range 97%
Linear Formula CH3OC6H4CH2CO2CH3
Name Note 97%
Refractive Index 1.5150 to 1.5170 (20°C, 589nm)
Density 1.1170g/mL
Formula Weight 180.20
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.