Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4'-methoxybiphenyl-4-carboxylate, 95%

$144.18 - $144.18

Chemical Identifiers

Chemical Name or Material Methyl 4'-methoxybiphenyl-4-carboxylate
CAS 729-17-9
Molecular Formula C15H14O3
Formula Weight 242.27g/mol
MDL Number MFCD01122234
Synonym 4'-Methoxybiphenyl-4-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5192406

 
alfa aesar™
H51924-06
Each for $144.18
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material Methyl 4'-methoxybiphenyl-4-carboxylate
CAS 729-17-9
Assay 95%
Molecular Formula C15H14O3
Formula Weight 242.27g/mol
MDL Number MFCD01122234
Synonym 4'-Methoxybiphenyl-4-carboxylic acid methyl ester