missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-Methoxyacetoacetate, TCI America™

Manufacturer:  TCI America M103125G

 View more versions of this product

Catalog No. M103125G



Specifications

Specifications

Methyl 4-Methoxyacetoacetate
Yellow
41051-15-4
MFCD00010183
QGBPKJFJAVDUNC-UHFFFAOYSA-N
methyl 4-methoxy-3-oxobutanoate
123500
Clear Liquid at 20°C
113°C
25g
C6H10O4
methyl 4-methoxyacetoacetate, methyl-4-methoxyacetoacetate, methyl 4-methoxy-3-oxobutyrate, butanoic acid, 4-methoxy-3-oxo-, methyl ester, methyl methoxyacetoacetate, 4-methoxyacetoacetic acid methyl ester, 4-methoxy-3-oxo-butyric acid methyl ester, pubchem10473, acmc-1aoxb, methyl4-methoxyacetoacetate
COCC(=O)CC(=O)OC
146.142
146.14
≥97.0% (GC)

Chemical Identifiers

41051-15-4
146.142
QGBPKJFJAVDUNC-UHFFFAOYSA-N
123500
COCC(=O)CC(=O)OC
C6H10O4
MFCD00010183
methyl 4-methoxyacetoacetate, methyl-4-methoxyacetoacetate, methyl 4-methoxy-3-oxobutyrate, butanoic acid, 4-methoxy-3-oxo-, methyl ester, methyl methoxyacetoacetate, 4-methoxyacetoacetic acid methyl ester, 4-methoxy-3-oxo-butyric acid methyl ester, pubchem10473, acmc-1aoxb, methyl4-methoxyacetoacetate
methyl 4-methoxy-3-oxobutanoate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents