Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-methoxy-3-nitrobenzoate, 97%

$149.92 - $557.50

Chemical Identifiers

Chemical Name or Material Methyl 4-methoxy-3-nitrobenzoate
CAS 40757-20-8
Molecular Formula C9H9NO5
Formula Weight 211.18
MDL Number MFCD00024213
Synonym 4-Methoxy-3-nitrobenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2524406

  SDS
alfa aesar™
B2524406
5g Each for $149.92

AAB2524414

  SDS
alfa aesar™
B2524414
25g Each for $557.50
Description & Specifications

Specifications

Formula Weight 211.18
Molecular Formula C9H9NO5
Quantity 5g
Synonym 4-Methoxy-3-nitrobenzoic acid methyl ester
Assay 97%
MDL Number MFCD00024213
Chemical Name or Material Methyl 4-methoxy-3-nitrobenzoate
Melting Point 109° to 112°C
CAS 40757-20-8