missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-Methoxy-2-methylbenzoate, TCI America™

$172.64 - $516.41

Chemical Identifiers

CAS 35598-05-1
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD06203918
InChI Key OGYAVWKYDVBIMW-UHFFFAOYSA-N
Synonym 4-Methoxy-2-methylbenzoic Acid Methyl Ester, Methyl 4-Methoxy-o-toluate, 4-Methoxy-o-toluic Acid Methyl Ester, Methyl 2-Methyl-p-anisate, 2-Methyl-p-anisic Acid Methyl Ester
PubChem CID 13257269
IUPAC Name methyl 4-methoxy-2-methylbenzoate
SMILES CC1=C(C=CC(=C1)OC)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M28891G View Documents TCI America
M28891G
1g Each for $172.64
 
M28895G View Documents TCI America
M28895G
5g Each for $516.41
 
Specifications

Chemical Identifiers

35598-05-1
180.203
OGYAVWKYDVBIMW-UHFFFAOYSA-N
13257269
CC1=C(C=CC(=C1)OC)C(=O)OC
C10H12O3
MFCD06203918
4-Methoxy-2-methylbenzoic Acid Methyl Ester, Methyl 4-Methoxy-o-toluate, 4-Methoxy-o-toluic Acid Methyl Ester, Methyl 2-Methyl-p-anisate, 2-Methyl-p-anisic Acid Methyl Ester
methyl 4-methoxy-2-methylbenzoate

Specifications

98°C
1g
C10H12O3
4-Methoxy-2-methylbenzoic Acid Methyl Ester, Methyl 4-Methoxy-o-toluate, 4-Methoxy-o-toluic Acid Methyl Ester, Methyl 2-Methyl-p-anisate, 2-Methyl-p-anisic Acid Methyl Ester
CC1=C(C=CC(=C1)OC)C(=O)OC
180.203
180.20
≥98.0% (GC)
Yellow
35598-05-1
MFCD06203918
OGYAVWKYDVBIMW-UHFFFAOYSA-N
methyl 4-methoxy-2-methylbenzoate
13257269
Clear Liquid at 20°C
Methyl 4-Methoxy-2-methylbenzoate
Safety and Handling

Safety and Handling

TSCA : No