Please login to your online account to display your discounted pricing

Methyl 4-iodobenzoate 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  414460250

 View more versions of this product

Catalog No. AC414460250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 262.04
Quantity 25g
Color Beige to pale pink
EINECS Number 210-597-1
TSCA TSCA
Melting Point 113° to 117°C
Packaging Glass bottle
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
Avoid release to the environment.
Infrared Spectrum Authentic
Chemical Name or Material Methyl 4-iodobenzoate
Assay Percent Range 98%
Molecular Formula C8H7IO2
Linear Formula CO2CH3C6H4I
MDL Number MFCD00016353
CAS 619-44-3
Health Hazard 2 GHS P Statement
Toxic to aquatic life with long lasting effects.
Name Note 98%

Specifications

Chemical Name or Material Methyl 4-iodobenzoate
Name Note 98%
CAS 619-44-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H7IO2
Linear Formula CO2CH3C6H4I
Formula Weight 262.04
MDL Number MFCD00016353
Accessories

Warning

GHS P Statement
Avoid release to the environment.

GHS P Statement
Toxic to aquatic life with long lasting effects.

SDS & Safety Documents