missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-hydroxyphenylacetate, 99%, ACROS Organics™

Manufacturer:  Acros Organics 208550250

 View more versions of this product

Catalog No. AC208550250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Methyl 4-hydroxyphenylacetate
Glass bottle
White to Yellow
25g
14199-15-6
C9H10O3
MFCD00002387
4-hydroxy-phenyl-acetic acid methyl ester
COC(=O)CC1=CC=C(C=C1)O
166.18
CHEBI:68078
Crystals
Authentic
162°C to 163°C (5.0mmHg)
55°C to 58°C
99%
99%
HOC6H4CH2CO2CH3
10, 191
XGDZEDRBLVIUMX-UHFFFAOYSA-N
methyl 2-(4-hydroxyphenyl)acetate
518900
166.18
≥98.5%

Chemical Identifiers

14199-15-6
166.18
XGDZEDRBLVIUMX-UHFFFAOYSA-N
518900
methyl 2-(4-hydroxyphenyl)acetate
C9H10O3
MFCD00002387
4-hydroxy-phenyl-acetic acid methyl ester
CHEBI:68078
COC(=O)CC1=CC=C(C=C1)O
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 238-050-2

TSCA : TSCA

SDS Documents