Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl (4-hydroxymethyl)benzoate, 99%

$77.15 - $301.35

Chemical Identifiers

Chemical Name or Material Methyl (4-hydroxymethyl)benzoate
CAS 6908-41-4
Molecular Formula C9H10O3
Formula Weight 166.18
MDL Number MFCD00009984
Synonym 4-(Hydroxymethyl)benzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2256406

  SDS
alfa aesar™
B2256406
5g Each for $77.15

AAB2256414

  SDS
alfa aesar™
B2256414
25g Each for $301.35
Description & Specifications

Specifications

Formula Weight 166.18
CAS 6908-41-4
Chemical Name or Material Methyl (4-hydroxymethyl)benzoate
Melting Point 46° to 52°C
Assay 99%
MDL Number MFCD00009984
Synonym 4-(Hydroxymethyl)benzoic acid methyl ester
Flash Point >110°C (230°F)
Molecular Formula C9H10O3
TSCA TSCA
Quantity 5g