missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-(hydroxymethyl)benzoate, 98%, ACROS Organics™

Chemical Identifiers

CAS 6908-41-4
Assay Percent Range 98%
Molecular Formula C9H10O3
Formula Weight 166.18
MDL Number MFCD00009984
Chemical Name or Material Methyl 4-(hydroxymethyl)benzoate, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC330840100 SDS Thermo Fisher Scientific
330840100
Glass bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6908-41-4
C9H10O3
MFCD00009984
≥97.5%
98%
166.18
Methyl 4-(hydroxymethyl)benzoate, 98%

Specifications

6908-41-4
Glass bottle
98%
166.18
White
10g
≥97.5%
Authentic
Powder or Cyrstals
C9H10O3
MFCD00009984
45° to 50°C
Methyl 4-(hydroxymethyl)benzoate, 98%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 230-017-0

TSCA : TSCA