Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-hydroxyindole-6-carboxylate, 97%

Chemical Identifiers

Chemical Name or Material Methyl 4-hydroxyindole-6-carboxylate
CAS 77140-48-8
Formula Weight 191.18
MDL Number MFCD04973985
Synonym 4-Hydroxyindole-6-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6339803

  SDS
alfa aesar™
H6339803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63398MD

  SDS
alfa aesar™
H63398MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.18
Chemical Name or Material Methyl 4-hydroxyindole-6-carboxylate
Physical Form Solid
Storage Note 1 2° to 8°C
Quantity 1g
Assay 97%
Color Pale pink
MDL Number MFCD04973985
CAS 77140-48-8
Synonym 4-Hydroxyindole-6-carboxylic acid methyl ester

Applications
Methyl 4-hydroxyindole-6-carboxylate is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water (2.5 g/L) (25°C).

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

2° to 8°C