Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4'-hydroxybiphenyl-3-carboxylate, 95%

Chemical Identifiers

Chemical Name or Material Methyl 4'-hydroxybiphenyl-3-carboxylate
CAS 192376-76-4
Molecular Formula C14H12O3
Formula Weight 228.25
MDL Number MFCD06409246
UN Number UN3077
Synonym 4'-Hydroxybiphenyl-3-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5177003

 
alfa aesar™
H5177003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5177006

 
alfa aesar™
H5177006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 228.25
Chemical Name or Material Methyl 4'-hydroxybiphenyl-3-carboxylate
Assay 95%
Molecular Formula C14H12O3
MDL Number MFCD06409246
CAS 192376-76-4
UN Number UN3077
Synonym 4'-Hydroxybiphenyl-3-carboxylic acid methyl ester
Health Hazard 1 Danger
Quantity 1g
Health Hazard 2 GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.