missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl (4-Hydroxybenzoyl)acetate, TCI America™

$357.50 - $1,216.14

Chemical Identifiers

CAS 32066-29-8
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00191516
InChI Key VZOFEVHSVUBEPH-UHFFFAOYSA-N
Synonym (4-Hydroxybenzoyl)acetic Acid Methyl Ester, Methyl 3-(4-Hydroxyphenyl)-3-oxopropionate, 3-(4-Hydroxyphenyl)-3-oxopropionic Acid Methyl Ester
PubChem CID 508465
IUPAC Name methyl 3-(4-hydroxyphenyl)-3-oxopropanoate
SMILES COC(=O)CC(=O)C1=CC=C(C=C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H08285G View Documents TCI America
H08285G
5g Each for $1,216.14
 
H08281G View Documents TCI America
H08281G
1g Each for $357.50
 
Specifications

Chemical Identifiers

32066-29-8
194.186
VZOFEVHSVUBEPH-UHFFFAOYSA-N
508465
COC(=O)CC(=O)C1=CC=C(C=C1)O
C10H10O4
MFCD00191516
(4-Hydroxybenzoyl)acetic Acid Methyl Ester, Methyl 3-(4-Hydroxyphenyl)-3-oxopropionate, 3-(4-Hydroxyphenyl)-3-oxopropionic Acid Methyl Ester
methyl 3-(4-hydroxyphenyl)-3-oxopropanoate

Specifications

White-Yellow
5g
C10H10O4
(4-Hydroxybenzoyl)acetic Acid Methyl Ester, Methyl 3-(4-Hydroxyphenyl)-3-oxopropionate, 3-(4-Hydroxyphenyl)-3-oxopropionic Acid Methyl Ester
COC(=O)CC(=O)C1=CC=C(C=C1)O
194.186
194.19
≥93.0% (GC)
90°C
32066-29-8
MFCD00191516
VZOFEVHSVUBEPH-UHFFFAOYSA-N
methyl 3-(4-hydroxyphenyl)-3-oxopropanoate
508465
Crystal-Powder at 20°C
Methyl (4-Hydroxybenzoyl)acetate
Safety and Handling

Safety and Handling

TSCA : No