Please login to your online account to display your discounted pricing

Methyl 4-hydroxybenzoate 99%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  126961000

 View more versions of this product

Catalog No. AC126961000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥98.5% (saponification)
Formula Weight 152.15
Quantity 100g
Color White to almost white
EINECS Number 202-785-7
TSCA TSCA
Melting Point 125° to 128°C
RTECS Number DH2450000
Packaging Plastic bottle
Residue after Ignition 0.1% max.
Synonym Methylparaben
Health Hazard 2 GHS P Statement Harmful to aquatic life with long lasting effects.
Infrared Spectrum Authentic
Chemical Name or Material Methyl 4-hydroxybenzoate
Name Note 99%
Assay Percent Range 99%
Molecular Formula C8H8O3
Linear Formula HOC6H4CO2CH3
MDL Number MFCD00002352
CAS 99-76-3
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Methyl 4-hydroxybenzoate
Name Note 99%
CAS 99-76-3
Assay ≥98.5% (saponification)
Assay Percent Range 99%
Molecular Formula C8H8O3
Linear Formula HOC6H4CO2CH3
Formula Weight 152.15
MDL Number MFCD00002352
Synonym Methylparaben
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement Harmful to aquatic life with long lasting effects.

SDS & Safety Documents