Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate, 99%


Quantity 1g

Manufacturer: alfa aesar™  L1462203

 View more versions of this product

Catalog No. AAL1462203

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 184.19
Assay 99%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Methyl 4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate
Molecular Formula C9H12O4
MDL Number MFCD01075661
CAS 39493-62-4
Beilstein 5258501
Synonym 5,6-Dihydroorsellinic acid methyl ester; 4-Hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate methyl ester
Melting Point 122° to 124°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning

Specifications

Chemical Name or Material Methyl 4-hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate
CAS 39493-62-4
Molecular Formula C9H12O4
Formula Weight 184.19
MDL Number MFCD01075661
Beilstein 5258501
Synonym 5,6-Dihydroorsellinic acid methyl ester; 4-Hydroxy-6-methyl-2-oxo-3-cyclohexene-1-carboxylate methyl ester
Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.