Learn More
Methyl 4-hydroxy-3-methoxycinnamate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2265704
Description
Methyl 4-hydroxy-3-methoxycinnamate has antioxidant activities that provide protection against inflammation and cancer. It has got the ability to cross cell membranes. It inhibits the release of pro-inflammatory cytokines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 4-hydroxy-3-methoxycinnamate has antioxidant activities that provide protection against inflammation and cancer. It has got the ability to cross cell membranes. It inhibits the release of pro-inflammatory cytokines.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Methyl 4-hydroxy-3-methoxycinnamate | |
2309-07-1 | |
C16H25ClN2O | |
oxymetazoline hydrochloride, oxymetazoline hcl, afrazine, ocuclear, lliadine, nasivin, nasivine, neonabel, nostrilla | |
BEEDODBODQVSIM-UHFFFAOYSA-N | |
6-tert-butyl-3-(4,5-dihydro-1H-imidazol-2-ylmethyl)-2,4-dimethylphenol;hydrochloride | |
66259 | |
208.22 |
62°C to 65°C | |
2 g | |
MFCD00017208 | |
Slightly soluble in water. | |
CC1=CC(=C(C(=C1CC2=NCCN2)C)O)C(C)(C)C.Cl | |
296.84 | |
CHEBI:7863 | |
99% |
Chemical Identifiers
2309-07-1 | |
MFCD00017208 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
RTECSNumber : GD9470000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only